Learn More
Medchemexpress LLC HY-10873 5mg Medchemexpress, N-Desethyl Sunitinib CAS:356068-97-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108735MG
Encompass_Preferred
Medchemexpress, HY-10873 5mg N-Desethyl Sunitinib CAS:356068-97-8 N-Desethyl Sunitinib is a metabolite of sunitinib. Sunitinib is a potent, ATP-competitive VEGFR, PDGFRβ and KIT inhibitor with Ki values of 2, 9, 17, 8 and 4 nM for VEGFR -1, -2, -3, PDGFRβ and KIT, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8622