Learn More
Medchemexpress LLC N-desethyl sunitinib hydrochloride | 99.1% | 406.88 | C20H24ClFN4O2 | 5 MG
SDP
Catalog No. 5000221431
Encompass_Preferred
N-desethyl sunitinib hydrochloride is the N-desethyl metabolite of the tyrosine kinase inhibitor sunitinib, supplied as a solid hydrochloride salt for laboratory research. It is provided in small, accurately weighed packages for use as a reference standard or in biochemical and cell-based kinase studies.
  • High purity (99.1%).
  • Supplied as a solid hydrochloride salt, yellow to brown.
  • Available in multiple small package sizes for research use.
  • Suitable as a metabolite reference in kinase inhibitor and pharmacokinetic studies.
  • Stable when stored sealed at 4°C; long-term storage in solvent at -80°C.
Catalog No. 50-002-21431 Supplier Medchemexpress LLC Supplier No. HY1388135MG
N-desethyl sunitinib hydrochloride is the N-desethyl metabolite of the tyrosine kinase inhibitor sunitinib, supplied as a solid hydrochloride salt for laboratory research. It is provided in small, accurately weighed packages for use as a reference standard or in biochemical and cell-based kinase studies.
  • High purity (99.1%).
  • Supplied as a solid hydrochloride salt, yellow to brown.
  • Available in multiple small package sizes for research use.
  • Suitable as a metabolite reference in kinase inhibitor and pharmacokinetic studies.
  • Stable when stored sealed at 4°C; long-term storage in solvent at -80°C.