Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-deshydroxyethyl dasatinib | 910297-51-7 | MFCD18252607 | 98.6% | 444.0 g/mol | C20H22ClN7OS | 10 MM 1 ML
SDP

Catalog No. 5000241506
Encompass_Preferred

N-Deshydroxyethyl dasatinib is a metabolite of the multi-kinase inhibitor dasatinib, provided as research-grade material for biochemical and cellular studies. It is supplied as both solid material and ready-to-use 10 mM solutions in DMSO for in vitro assays and method development.

  • Purity 98.58%
  • Available as 10 mM solution in DMSO (1 mL)
  • Also available as solid in multiple pack sizes (5-100 mg)
  • Molecular weight 444.0 g/mol
  • Chemical formula C20H22ClN7OS
  • Intended for research use only

Catalog No. 50-002-41506 Supplier Medchemexpress LLC Supplier No. HY10744710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Deshydroxyethyl dasatinib is a metabolite of the multi-kinase inhibitor dasatinib, provided as research-grade material for biochemical and cellular studies. It is supplied as both solid material and ready-to-use 10 mM solutions in DMSO for in vitro assays and method development.

  • Purity 98.58%
  • Available as 10 mM solution in DMSO (1 mL)
  • Also available as solid in multiple pack sizes (5-100 mg)
  • Molecular weight 444.0 g/mol
  • Chemical formula C20H22ClN7OS
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.