Learn More
Medchemexpress LLC N-desmethyl apalutamide | 99.7% | 463.41 g/mol | C20H13F4N5O2S | 1mg
SDP
Catalog No. 5000270508
Encompass_Preferred
N-Desmethyl apalutamide-d4 is the deuterium-labeled N-Desmethyl-Apalutamide (HY-135331) N-Desmethyl Apalutamide is an active metabolite of Apalutamide N-Desmethyl Apalutamide is a less potent antagonist of the androgen receptor and is responsible for one-third of the activity of Apalutamide The formation of N-Desmethyl Apalutamide mediated predominantly by CYP2C8 and CYP3A4 N-Desmethyl Apalutamide is moderate to strong CYP3A4 and CYP2B6 inducer and has an excellent plasma-proteins bound concentration[1][2][3]
Catalog No. 50-002-70508 Supplier Medchemexpress LLC Supplier No. HYW7541511MG
N-Desmethyl apalutamide-d4 is the deuterium-labeled N-Desmethyl-Apalutamide (HY-135331) N-Desmethyl Apalutamide is an active metabolite of Apalutamide N-Desmethyl Apalutamide is a less potent antagonist of the androgen receptor and is responsible for one-third of the activity of Apalutamide The formation of N-Desmethyl Apalutamide mediated predominantly by CYP2C8 and CYP3A4 N-Desmethyl Apalutamide is moderate to strong CYP3A4 and CYP2B6 inducer and has an excellent plasma-proteins bound concentration[1][2][3]