Learn More
Medchemexpress LLC N-desmethyl apalutamide | 1332391-11-3 | MFCD32858271 | 99.7% | 463.41 g/mol | C20H13F4N5O2S | 5 MG
SDP
Catalog No. 5000222898
Encompass_Preferred
N-desmethyl apalutamide is the major active metabolite of apalutamide that functions as a less potent androgen receptor antagonist and a moderate-strong inducer of CYP3A4 and CYP2B6. Supplied as a white to off-white solid for research use, it is characterized by high purity and defined storage conditions.
  • Active metabolite of apalutamide with androgen receptor antagonist activity.
  • Moderate to strong inducer of CYP3A4 and CYP2B6.
  • High purity (99.7%).
  • Molecular formula C20H13F4N5O2S and molecular weight 463.41 g/mol.
  • White to off-white solid suitable for analytical and pharmacology studies.
  • Specified storage conditions for powder and solutions to maintain stability.
Catalog No. 50-002-22898 Supplier Medchemexpress LLC Supplier No. HY1353315MG
N-desmethyl apalutamide is the major active metabolite of apalutamide that functions as a less potent androgen receptor antagonist and a moderate-strong inducer of CYP3A4 and CYP2B6. Supplied as a white to off-white solid for research use, it is characterized by high purity and defined storage conditions.
  • Active metabolite of apalutamide with androgen receptor antagonist activity.
  • Moderate to strong inducer of CYP3A4 and CYP2B6.
  • High purity (99.7%).
  • Molecular formula C20H13F4N5O2S and molecular weight 463.41 g/mol.
  • White to off-white solid suitable for analytical and pharmacology studies.
  • Specified storage conditions for powder and solutions to maintain stability.