Learn More
TARGETMOL CHEMICALS INC N-Desmethyl-Apalutamide 1MG
SDP
Catalog No. 502879541
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.N-Desmethyl-Apalutamide is a less potent antagonist of the androgen receptor, is an active Apalutamide metabolite. Purity 97.83%
Catalog No. 50-287-9541 Supplier TARGETMOL CHEMICALS INC Supplier No. T121461MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.N-Desmethyl-Apalutamide is a less potent antagonist of the androgen receptor, is an active Apalutamide metabolite. Purity 97.83%