Learn More
Cayman Chemical N-desmethyl Azelastine, Purity: ≥98%, CAS: 47491-38-3, 5 mg
SDP
Catalog No. 502913215
Encompass_Preferred
An active metabolite of azelastine; inhibits acetylcholine-induced depolarization and contractions in isolated human tracheal smooth muscle at 1 µM; inhibits transport of daunorubicin and digoxin in LLC-GA5-CoL150 cells overexpressing P-gp (IC50s = 11.8 and 41.8 µM, respectively)
Catalog No. 50-291-3215 Supplier Cayman Chemical Supplier No. 354295 MG
An active metabolite of azelastine; inhibits acetylcholine-induced depolarization and contractions in isolated human tracheal smooth muscle at 1 µM; inhibits transport of daunorubicin and digoxin in LLC-GA5-CoL150 cells overexpressing P-gp (IC50s = 11.8 and 41.8 µM, respectively)