Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Desmethyl Bedaquiline-d6 | 2271264-26-5 | 96.39% | C31H23D6BrN2O2 | 1 MG
SDP

Catalog No. 5000354568
Encompass_Preferred

N-Desmethyl Bedaquiline-d6 is a deuterium labeled compound designed for use as a tracer or internal standard in quantitative analysis methods such as NMR, GC-MS, and LC-MS. Its deuteration may impact the pharmacokinetic and metabolic profiles of drugs.

  • Used as a tracer
  • Internal standard for quantitative analysis
  • Stable heavy isotope
  • Molecular weight: 547.52
  • Appearance: White to off-white solid
  • For research use only

Catalog No. 50-003-54568 Supplier Medchemexpress LLC Supplier No. HY143922S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desmethyl Bedaquiline-d6 is a deuterium labeled compound designed for use as a tracer or internal standard in quantitative analysis methods such as NMR, GC-MS, and LC-MS. Its deuteration may impact the pharmacokinetic and metabolic profiles of drugs.

  • Used as a tracer
  • Internal standard for quantitative analysis
  • Stable heavy isotope
  • Molecular weight: 547.52
  • Appearance: White to off-white solid
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.