Learn More
Medchemexpress LLC N-Desmethyl citalopram-d4 oxalate | 2747915-54-2 | C21H17D4FN2O5 | 1 MG
SDP
Catalog No. 5000355649
Encompass_Preferred
N-Desmethyl citalopram-d4 (oxalate) is a deuterium-labeled N-Desmethyl citalopram oxalate. It can be utilized as a tracer or internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules to aid quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
  • Features stable heavy isotopes for drug development quantitation.
  • Potential to influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-55649 Supplier Medchemexpress LLC Supplier No. HY143931S1MG
N-Desmethyl citalopram-d4 (oxalate) is a deuterium-labeled N-Desmethyl citalopram oxalate. It can be utilized as a tracer or internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules to aid quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
  • Features stable heavy isotopes for drug development quantitation.
  • Potential to influence pharmacokinetic and metabolic profiles of drugs.