Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-desmethyl clomipramine-d3 (hydrochloride) | 1189971-04-7 | MFCD28899247 | 99.8% | 340.31 | C18H19D3Cl2N2 | 500UG
SDP

Catalog No. 5000236791
Encompass_Preferred

N-desmethyl clomipramine-d3 hydrochloride is a deuterium-labeled analytical standard intended for use as an internal standard in quantitative mass spectrometry and isotope-dilution assays. It is supplied as the hydrochloride salt and is intended for analytical and research applications requiring isotopically labeled reference material.

  • Deuterium-labeled internal standard for isotope-dilution mass spectrometry.
  • Hydrochloride salt form for improved stability and handling.
  • Purity 99.8% as reported on the product specification.
  • Molecular weight 340.31 g/mol.
  • Chemical formula C18H19D3Cl2N2.
  • Available in low microgram to milligram pack sizes, including 500 μg.

Catalog No. 50-002-36791 Supplier Medchemexpress LLC Supplier No. HY12388AS500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-desmethyl clomipramine-d3 hydrochloride is a deuterium-labeled analytical standard intended for use as an internal standard in quantitative mass spectrometry and isotope-dilution assays. It is supplied as the hydrochloride salt and is intended for analytical and research applications requiring isotopically labeled reference material.

  • Deuterium-labeled internal standard for isotope-dilution mass spectrometry.
  • Hydrochloride salt form for improved stability and handling.
  • Purity 99.8% as reported on the product specification.
  • Molecular weight 340.31 g/mol.
  • Chemical formula C18H19D3Cl2N2.
  • Available in low microgram to milligram pack sizes, including 500 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.