Learn More
Medchemexpress LLC N-desmethyl clomipramine-d3 hydrochloride | 1189971-04-7 | MFCD28899247 | 99.8% | 340.31 g/mol | C18H19D3Cl2N2 | 1MG
SDP
Catalog No. 5000236989
Encompass_Preferred
N-Desmethyl clomipramine-d3 hydrochloride is the deuterium-labeled hydrochloride salt of N-desmethyl clomipramine (norclomipramine), supplied for research and analytical applications. It is intended primarily as an internal standard for quantitative mass spectrometry and pharmacokinetic studies involving clomipramine metabolism.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for pharmacokinetic and metabolic studies.
  • High chemical purity, 99.8%.
  • Provided as a hydrochloride salt for improved solubility.
  • Available in small analytical pack sizes for research use.
Catalog No. 50-002-36989 Supplier Medchemexpress LLC Supplier No. HY12388AS1MG
N-Desmethyl clomipramine-d3 hydrochloride is the deuterium-labeled hydrochloride salt of N-desmethyl clomipramine (norclomipramine), supplied for research and analytical applications. It is intended primarily as an internal standard for quantitative mass spectrometry and pharmacokinetic studies involving clomipramine metabolism.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Suitable for pharmacokinetic and metabolic studies.
  • High chemical purity, 99.8%.
  • Provided as a hydrochloride salt for improved solubility.
  • Available in small analytical pack sizes for research use.