Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-desmethyl clomipramine hydrochloride | 29854-14-6 | MFCD00792880 | 100.0% | 337.29 | C18H22Cl2N2 | 10MM 1ML
SDP

Catalog No. 5000236790
Encompass_Preferred

N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl metabolite of the tricyclic antidepressant clomipramine. Supplied as the hydrochloride salt for research use, the compound is offered in a 10 mM solution in DMSO (1 mL) and in solid quantities for analytical, metabolic, and pharmacological studies.

  • Primary N-desmethyl metabolite used as a reference standard.
  • Available as a 10 mM solution in DMSO (1 mL) and multiple solid pack sizes.
  • High purity (99.95%) suitable for analytical and in vitro applications.
  • Molecular weight 337.29 and formula C18H22Cl2N2 for accurate calculations.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-36790 Supplier Medchemexpress LLC Supplier No. HY12388A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl metabolite of the tricyclic antidepressant clomipramine. Supplied as the hydrochloride salt for research use, the compound is offered in a 10 mM solution in DMSO (1 mL) and in solid quantities for analytical, metabolic, and pharmacological studies.

  • Primary N-desmethyl metabolite used as a reference standard.
  • Available as a 10 mM solution in DMSO (1 mL) and multiple solid pack sizes.
  • High purity (99.95%) suitable for analytical and in vitro applications.
  • Molecular weight 337.29 and formula C18H22Cl2N2 for accurate calculations.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.