Learn More
Medchemexpress LLC N-desmethyl clomipramine hydrochloride | 29854-14-6 | MFCD00792880 | 99.95% | 337.29 | C18H22Cl2N2 | 50MG
SDP
Catalog No. 5000235157
Encompass_Preferred
N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl plasma metabolite of the tricyclic antidepressant clomipramine, supplied as a research-grade chemical standard for analytical, metabolic, and neurological studies. It is provided as a solid (white to pink) with a high stated purity and is offered in small laboratory pack sizes and solution formats.
  • Primary N-desmethyl metabolite of clomipramine suitable for metabolite studies.
  • High stated purity: 99.95%.
  • Available as a solid and as a 10 mM solution in DMSO.
  • White to pink solid appearance for easy visual identification.
  • Suitable as an analytical reference standard for neurological and metabolic research.
  • Offered in multiple small pack sizes for lab-scale use.
Catalog No. 50-002-35157 Supplier Medchemexpress LLC Supplier No. HY12388A50MG
N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl plasma metabolite of the tricyclic antidepressant clomipramine, supplied as a research-grade chemical standard for analytical, metabolic, and neurological studies. It is provided as a solid (white to pink) with a high stated purity and is offered in small laboratory pack sizes and solution formats.
  • Primary N-desmethyl metabolite of clomipramine suitable for metabolite studies.
  • High stated purity: 99.95%.
  • Available as a solid and as a 10 mM solution in DMSO.
  • White to pink solid appearance for easy visual identification.
  • Suitable as an analytical reference standard for neurological and metabolic research.
  • Offered in multiple small pack sizes for lab-scale use.