Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-desmethyl clomipramine hydrochloride | 29854-14-6 | MFCD00792880 | 100.0% | 337.29 g/mol | C18H22Cl2N2 | 100MG
SDP

Catalog No. 5000235669
Encompass_Preferred

N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl metabolite of clomipramine, supplied as the hydrochloride salt for use as an analytical reference standard in research on antidepressant metabolism and pharmacology. The product is provided as a solid powder in a range of pack sizes for analytical assays, method development, and reference material applications.

  • Hydrochloride salt form
  • Molecular weight 337.29 g/mol
  • Chemical formula C18H22Cl2N2
  • Provided as a solid powder
  • Available in multiple pack sizes including 100 mg
  • Intended for analytical and research use

Catalog No. 50-002-35669 Supplier Medchemexpress LLC Supplier No. HY12388A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl metabolite of clomipramine, supplied as the hydrochloride salt for use as an analytical reference standard in research on antidepressant metabolism and pharmacology. The product is provided as a solid powder in a range of pack sizes for analytical assays, method development, and reference material applications.

  • Hydrochloride salt form
  • Molecular weight 337.29 g/mol
  • Chemical formula C18H22Cl2N2
  • Provided as a solid powder
  • Available in multiple pack sizes including 100 mg
  • Intended for analytical and research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.