Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-desmethyl clomipramine hydrochloride | 29854-14-6 | MFCD00792880 | 100.0% | 337.29 g/mol | C18H22Cl2N2 | 10MG
SDP

Catalog No. 5000236246
Encompass_Preferred

N-Desmethyl clomipramine hydrochloride is the primary plasma N-desmethyl metabolite of clomipramine, supplied as the hydrochloride salt for research use in pharmacology, metabolism, and analytical studies. Provided as a solid reference material with defined storage guidance for solid and solution preparations.

  • CAS number 29854-14-6.
  • Molecular formula C18H22Cl2N2.
  • Molecular weight 337.29 g/mol.
  • Appearance: solid, white to pink.
  • High purity suitable for analytical and reference use.
  • Storage: solid at 4°C; solutions recommended at -80°C or -20°C.
  • Typical applications: pharmacology, metabolism, and analytical studies.

Catalog No. 50-002-36246 Supplier Medchemexpress LLC Supplier No. HY12388A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desmethyl clomipramine hydrochloride is the primary plasma N-desmethyl metabolite of clomipramine, supplied as the hydrochloride salt for research use in pharmacology, metabolism, and analytical studies. Provided as a solid reference material with defined storage guidance for solid and solution preparations.

  • CAS number 29854-14-6.
  • Molecular formula C18H22Cl2N2.
  • Molecular weight 337.29 g/mol.
  • Appearance: solid, white to pink.
  • High purity suitable for analytical and reference use.
  • Storage: solid at 4°C; solutions recommended at -80°C or -20°C.
  • Typical applications: pharmacology, metabolism, and analytical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.