Learn More
Medchemexpress LLC N-desmethyl clomipramine hydrochloride | 29854-14-6 | MFCD00792880 | 100.0% | 337.29 g/mol | C18H22Cl2N2 | 25MG
SDP
Catalog No. 5000237020
Encompass_Preferred
N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl plasma metabolite of the tricyclic antidepressant clomipramine, supplied for research use in neuroscience, pharmacokinetics, and drug metabolism studies. The compound is provided as a solid and is characterized by the chemical formula C18H22Cl2N2 and molecular weight 337.29 g/mol.
  • High purity suitable for analytical and reference standard use.
  • Available in small solid pack sizes, including 25 mg, and in solution formats.
  • Characterized by supporting documents such as datasheet, COA, SDS, and analytical spectra.
  • Applicable for metabolic, pharmacokinetic, and neuropharmacology research.
  • Stable solid form with storage and handling guidance provided in the SDS.
Catalog No. 50-002-37020 Supplier Medchemexpress LLC Supplier No. HY12388A25MG
N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl plasma metabolite of the tricyclic antidepressant clomipramine, supplied for research use in neuroscience, pharmacokinetics, and drug metabolism studies. The compound is provided as a solid and is characterized by the chemical formula C18H22Cl2N2 and molecular weight 337.29 g/mol.
  • High purity suitable for analytical and reference standard use.
  • Available in small solid pack sizes, including 25 mg, and in solution formats.
  • Characterized by supporting documents such as datasheet, COA, SDS, and analytical spectra.
  • Applicable for metabolic, pharmacokinetic, and neuropharmacology research.
  • Stable solid form with storage and handling guidance provided in the SDS.