Learn More
Medchemexpress LLC N-desmethyl clomipramine hydrochloride | 29854-14-6 | MFCD00792880 | 100.0% | 337.29 g/mol | C18H22Cl2N2 | 5MG
SDP
Catalog No. 5000236108
Encompass_Preferred
N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl metabolite of the tricyclic antidepressant clomipramine. Supplied as the hydrochloride salt for research use, it is used as an analytical reference standard and for biochemical or pharmacological studies. The compound is a solid (white to pink), with formula C18H22Cl2N2, molecular weight 337.29 g/mol, and CAS 29854-14-6. Packaging includes mg-scale quantities suitable for analytical workflows.
  • Primary N-desmethyl metabolite of clomipramine
  • Hydrochloride salt supplied as a solid, white to pink
  • Suitable as an analytical reference standard for assays
  • Available in small-scale packaging for laboratory use
  • High purity appropriate for analytical and research applications
Catalog No. 50-002-36108 Supplier Medchemexpress LLC Supplier No. HY12388A5MG
N-Desmethyl clomipramine hydrochloride is the primary N-desmethyl metabolite of the tricyclic antidepressant clomipramine. Supplied as the hydrochloride salt for research use, it is used as an analytical reference standard and for biochemical or pharmacological studies. The compound is a solid (white to pink), with formula C18H22Cl2N2, molecular weight 337.29 g/mol, and CAS 29854-14-6. Packaging includes mg-scale quantities suitable for analytical workflows.
  • Primary N-desmethyl metabolite of clomipramine
  • Hydrochloride salt supplied as a solid, white to pink
  • Suitable as an analytical reference standard for assays
  • Available in small-scale packaging for laboratory use
  • High purity appropriate for analytical and research applications