Learn More
Enzo Life Sciences N-Desmethyl Clozapine (25mg). CAS: 6104-71-8
SDP
Supplier:  Enzo Life Sciences BMLNS1060025
Encompass_Preferred
Major metabolite of clozapine. Potent serotonin 5-HT1c receptor antagonist. Alternative name: Norclozapine. Purity: ≥99% (HPLC). Solubility: Soluble in ethanol (25 mg/ml) or DMSO (>44 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1203