Learn More
Abcam N-Desmethyl clozapine (Normethylclozapine), Clozapine metabolite. M1 agonist. D2/D3 agonist., 10MG
Catalog No. 502707923
Encompass_Preferred
MW 312.8 Da, Purity >99%. Brain penetrant metabolite of the atypical antipsychotic clozapine. Broad range of actions. Potent allosteric M1 partial agonist (IC50 = 55 nM for inhibition of [3H]-NMS binding). D2/D3 partial agonist. Also displays affinity at 5-HT1 and 5-HT2.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7923 Supplier Abcam Supplier No. AB12002610MG
MW 312.8 Da, Purity >99%. Brain penetrant metabolite of the atypical antipsychotic clozapine. Broad range of actions. Potent allosteric M1 partial agonist (IC50 = 55 nM for inhibition of [3H]-NMS binding). D2/D3 partial agonist. Also displays affinity at 5-HT1 and 5-HT2.
The product is subject to the following: Abcam Restricted Use Statement