Learn More
Medchemexpress LLC N-desmethyl diltiazem hydrochloride | 130606-60-9 | 10mg
SDP
Catalog No. 5000269948
Encompass_Preferred
N-Desmethyl diltiazem hydrochloride is a substrate for CYP3A7 metabolism which is the predominant fetal form and is minimally expressed in adults[1]
Catalog No. 50-002-69948 Supplier Medchemexpress LLC Supplier No. HYZ803310MG
N-Desmethyl diltiazem hydrochloride is a substrate for CYP3A7 metabolism which is the predominant fetal form and is minimally expressed in adults[1]