Learn More
Medchemexpress LLC N-desmethyl Enzalutamide | 1242137-16-1 | 99.6% | C20H14F4N4O2S | 1 ML
SDP
Catalog No. 5000433579
Encompass_Preferred
N-desmethyl Enzalutamide is the active metabolite of Enzalutamide. It demonstrates primary and secondary pharmacodynamics of similar potency and circulates at approximately the same plasma concentrations as enzalutamide.
  • Active metabolite of enzalutamide
  • Demonstrates similar potency to enzalutamide
  • Circulates at approximately the same plasma concentrations as enzalutamide
  • For research use only
Catalog No. 50-004-33579 Supplier Medchemexpress LLC Supplier No. HY70002A10MM/1ML
N-desmethyl Enzalutamide is the active metabolite of Enzalutamide. It demonstrates primary and secondary pharmacodynamics of similar potency and circulates at approximately the same plasma concentrations as enzalutamide.
  • Active metabolite of enzalutamide
  • Demonstrates similar potency to enzalutamide
  • Circulates at approximately the same plasma concentrations as enzalutamide
  • For research use only