Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Desmethyl Galanthamine | 41303-74-6 | 1 MG
SDP

Catalog No. 5000417232
Encompass_Preferred

N-Desmethyl Galanthamine (N-Norgalanthamine) is a metabolite of Galanthamine. It is an EeAChE inhibitor with an IC50 of 2.76 μM. This product can be used in the research of Alzheimer's disease.

  • Metabolite of Galanthamine.
  • EeAChE inhibitor (IC50: 2.76 μM).
  • Applicable in Alzheimer's disease research.
  • Purity: 99.60%.
  • Appearance: Solid, White to off-white.
  • Molecular weight: 273.33.
  • Formula: C16H19NO3.

Catalog No. 50-004-17232 Supplier Medchemexpress LLC Supplier No. HYN76121MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desmethyl Galanthamine (N-Norgalanthamine) is a metabolite of Galanthamine. It is an EeAChE inhibitor with an IC50 of 2.76 μM. This product can be used in the research of Alzheimer's disease.

  • Metabolite of Galanthamine.
  • EeAChE inhibitor (IC50: 2.76 μM).
  • Applicable in Alzheimer's disease research.
  • Purity: 99.60%.
  • Appearance: Solid, White to off-white.
  • Molecular weight: 273.33.
  • Formula: C16H19NO3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.