Learn More
Cayman Chemical N-desmethyl ImatInIb 10mg
SDP
Catalog No. 502967722
Encompass_Preferred
A major active metabolite of imatinib; formed through demethylation of imatinib by CYP3A4; has the same in vitro potency at Bcr-ABL kinase as imatinib (IC50 = 38 nM for both) but is only present in plasma at 10-15% of the levels of imatinib
Catalog No. 50-296-7722 Supplier Cayman Chemical Supplier No. 1694710 MG
A major active metabolite of imatinib; formed through demethylation of imatinib by CYP3A4; has the same in vitro potency at Bcr-ABL kinase as imatinib (IC50 = 38 nM for both) but is only present in plasma at 10-15% of the levels of imatinib