Learn More
Cayman Chemical N-desmethyl RosIglItazon 10mg
SDP
Supplier:  Cayman Chemical 2210310 MG
Encompass_Preferred
A major metabolite of rosiglitazone; formed through CYP2C8 metabolism of rosiglitazone
Catalog No. 50-298-7721