Learn More
Cayman Chemical N-desmethyl Sildenafil, Purity: ≥98%, CAS: 139755-82-1, 10 mg
SDP
Catalog No. 502967700
Encompass_Preferred
A major metabolite of sildenafil; formed via oxidative metabolism of sildenafil by CYP3A4, CYP3A5, and CYP3A7
Catalog No. 50-296-7700 Supplier Cayman Chemical Supplier No. 1820810 MG
A major metabolite of sildenafil; formed via oxidative metabolism of sildenafil by CYP3A4, CYP3A5, and CYP3A7