Learn More
Apexbio Technology LLC N-Desmethyl Sildenafil 139755-82-1 5mg
SDP
Catalog No. 5000094573
Encompass_Preferred
N-Desmethyl Sildenafil (CAS 139755-82-1) is a pharmacologically active metabolite of sildenafil It acts as a selective inhibitor of phosphodiesterase type 5 (PDE5) modulating intracellular signaling pathways by preventing the degradation of cyclic guanosine monophosphate (cGMP) Experimental studies have demonstrated its inhibitory activity toward PDE5 implicating a role in the modulation of vascular and smooth muscle functions N-Desmethyl Sildenafil is utilized in biomedical research to study PDE5-related cellular processes and to explore potential therapeutic strategies targeting this enzyme
Catalog No. 50-000-94573 Supplier Apexbio Technology LLC Supplier No. A46425
N-Desmethyl Sildenafil (CAS 139755-82-1) is a pharmacologically active metabolite of sildenafil It acts as a selective inhibitor of phosphodiesterase type 5 (PDE5) modulating intracellular signaling pathways by preventing the degradation of cyclic guanosine monophosphate (cGMP) Experimental studies have demonstrated its inhibitory activity toward PDE5 implicating a role in the modulation of vascular and smooth muscle functions N-Desmethyl Sildenafil is utilized in biomedical research to study PDE5-related cellular processes and to explore potential therapeutic strategies targeting this enzyme