Learn More
Medchemexpress LLC N-desmethyl sildenafil | 139755-82-1 | MFCD00908398 | 99.5% | 460.55 | C21H28N6O4S | 10 MM 1 ML
SDP
Catalog No. 5000228470
Encompass_Preferred
N-Desmethyl sildenafil is the primary N-desalkylated metabolite of sildenafil, used as a biochemical reagent and reference standard in pharmacology and metabolic studies of phosphodiesterase (PDE) activity. It is provided in solution and solid formats for analytical and experimental workflows.
  • Available as a 10 mM solution in DMSO and as solid quantities for experimental flexibility.
  • High purity suitable for analytical, pharmacological, and metabolite identification studies.
  • Applicable as a reference standard in metabolic profiling and mass spectrometry workflows.
  • Solution format enables ready-to-use dosing for in vitro assays; solid format supports long-term storage.
Catalog No. 50-002-28470 Supplier Medchemexpress LLC Supplier No. HY11760510MM 1ML
N-Desmethyl sildenafil is the primary N-desalkylated metabolite of sildenafil, used as a biochemical reagent and reference standard in pharmacology and metabolic studies of phosphodiesterase (PDE) activity. It is provided in solution and solid formats for analytical and experimental workflows.
  • Available as a 10 mM solution in DMSO and as solid quantities for experimental flexibility.
  • High purity suitable for analytical, pharmacological, and metabolite identification studies.
  • Applicable as a reference standard in metabolic profiling and mass spectrometry workflows.
  • Solution format enables ready-to-use dosing for in vitro assays; solid format supports long-term storage.