Learn More
Medchemexpress LLC N-desmethyl sildenafil | 139755-82-1 | MFCD00908398 | 99.5% | 460.55 g/mol | C21H28N6O4S | 10 MG
SDP
Catalog No. 5000226588
Encompass_Preferred
N-Desmethyl sildenafil is the major N-desmethyl metabolite of sildenafil and is supplied as a high-purity analytical standard for research and analytical applications related to phosphodiesterase type 5 (PDE5) activity and metabolism. The compound is characterized by a defined molecular formula and molecular weight, and is provided in small milligram pack sizes for reference and trace-level analyses.
  • High purity (≈99.5%) suitable for analytical and reference use.
  • Intended for analytical, reference, and research applications related to PDE5 activity.
  • Available in small milligram pack sizes for trace-level studies.
  • Molecular formula C21H28N6O4S; molecular weight approximately 460.55 g/mol.
  • Store powder at -20°C for long-term stability; follow solvent storage guidelines for solutions.
Catalog No. 50-002-26588 Supplier Medchemexpress LLC Supplier No. HY11760510MG
N-Desmethyl sildenafil is the major N-desmethyl metabolite of sildenafil and is supplied as a high-purity analytical standard for research and analytical applications related to phosphodiesterase type 5 (PDE5) activity and metabolism. The compound is characterized by a defined molecular formula and molecular weight, and is provided in small milligram pack sizes for reference and trace-level analyses.
  • High purity (≈99.5%) suitable for analytical and reference use.
  • Intended for analytical, reference, and research applications related to PDE5 activity.
  • Available in small milligram pack sizes for trace-level studies.
  • Molecular formula C21H28N6O4S; molecular weight approximately 460.55 g/mol.
  • Store powder at -20°C for long-term stability; follow solvent storage guidelines for solutions.