Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-desmethyl sildenafil-d8 | 1185168-06-2 | 99.3% | 468.60 | C21H20D8N6O4S | 1 MG
SDP

Catalog No. 5000227851
Encompass_Preferred

N-Desmethyl Sildenafil-d8 is the deuterium-labeled form of N-desmethyl sildenafil supplied as a high-purity analytical standard for LC-MS/MS, pharmacokinetic, and metabolic studies. It provides a stable isotope internal standard for accurate quantitation and method development.

  • Deuterium-labeled internal standard for mass spectrometry
  • High purity (99.3%) for reliable quantitation
  • Molecular formula C21H20D8N6O4S; molecular weight 468.60
  • Available in small quantities suitable for analytical use (1 mg, 5 mg)
  • Stable isotope reduces matrix effects and improves accuracy

Catalog No. 50-002-27851 Supplier Medchemexpress LLC Supplier No. HY117605S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desmethyl Sildenafil-d8 is the deuterium-labeled form of N-desmethyl sildenafil supplied as a high-purity analytical standard for LC-MS/MS, pharmacokinetic, and metabolic studies. It provides a stable isotope internal standard for accurate quantitation and method development.

  • Deuterium-labeled internal standard for mass spectrometry
  • High purity (99.3%) for reliable quantitation
  • Molecular formula C21H20D8N6O4S; molecular weight 468.60
  • Available in small quantities suitable for analytical use (1 mg, 5 mg)
  • Stable isotope reduces matrix effects and improves accuracy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.