Learn More
Medchemexpress LLC N-desmethyl sildenafil-d8 | 1185168-06-2 | MFCD11977920 | 99.3% | 468.60 g/mol | C21H20D8N6O4S | 5 MG
SDP
Catalog No. 5000227679
Encompass_Preferred
N-Desmethyl sildenafil-d8 is a deuterium-labeled analytical standard of the N-desmethyl metabolite of sildenafil, provided in milligram quantities for research and quantitative analysis. It is intended for use as an internal standard in mass-spectrometry, pharmacokinetic, and metabolic studies, and is supplied with recommended storage conditions to preserve isotopic integrity.
  • Deuterium-labeled internal standard for quantitative LC-MS/MS analysis.
  • High purity suitable for analytical and pharmacokinetic studies.
  • Supplied in small milligram amounts for precise measurements.
  • Stable under recommended storage to maintain isotopic composition.
  • Appropriate as a reference material for metabolite identification.
Catalog No. 50-002-27679 Supplier Medchemexpress LLC Supplier No. HY117605S5MG
N-Desmethyl sildenafil-d8 is a deuterium-labeled analytical standard of the N-desmethyl metabolite of sildenafil, provided in milligram quantities for research and quantitative analysis. It is intended for use as an internal standard in mass-spectrometry, pharmacokinetic, and metabolic studies, and is supplied with recommended storage conditions to preserve isotopic integrity.
  • Deuterium-labeled internal standard for quantitative LC-MS/MS analysis.
  • High purity suitable for analytical and pharmacokinetic studies.
  • Supplied in small milligram amounts for precise measurements.
  • Stable under recommended storage to maintain isotopic composition.
  • Appropriate as a reference material for metabolite identification.