Learn More
Medchemexpress LLC N-desmethyl venlafaxine hydrochloride | 93413-90-2 | MFCD18711349 | 99.9% | C16H26ClNO2 | 10MG
SDP
Catalog No. 5000260670
Encompass_Preferred
Wy-45494 hydrochloride (N-desmethyl venlafaxine hydrochloride) is an analytical reference standard of the major active metabolite of venlafaxine supplied as the hydrochloride salt. It is provided with documented characterization and high purity for use in analytical method development, assay validation, and pharmacological research.
  • High purity suitable for analytical applications.
  • Characterized molecular formula and molecular weight for identification.
  • Suitable as a reference standard for chromatography and mass spectrometry.
  • Demonstrated activity inhibiting norepinephrine and serotonin reuptake in vitro.
  • Provided with a certificate of analysis for traceability.
  • Intended for research use only.
Catalog No. 50-002-60670 Supplier Medchemexpress LLC Supplier No. HY13152710MG
Wy-45494 hydrochloride (N-desmethyl venlafaxine hydrochloride) is an analytical reference standard of the major active metabolite of venlafaxine supplied as the hydrochloride salt. It is provided with documented characterization and high purity for use in analytical method development, assay validation, and pharmacological research.
  • High purity suitable for analytical applications.
  • Characterized molecular formula and molecular weight for identification.
  • Suitable as a reference standard for chromatography and mass spectrometry.
  • Demonstrated activity inhibiting norepinephrine and serotonin reuptake in vitro.
  • Provided with a certificate of analysis for traceability.
  • Intended for research use only.