Learn More
Cayman Chemical N-DesmethylclozapIn 10mg
SDP
Catalog No. 502936972
Encompass_Preferred
An active metabolite of clozapine; originally described as an antagonist of 5-HT2C receptors (IC50 = 7.1 nM in rat choroid plexus); and later as an inverse agonist using human recombinant receptors; an antagonist at dopamine D4 receptors, an agonist at δ-opioid receptors, and a partial agonist at dopamine D2 and D3, M1 muscarinic acetylcholine, and 5-HT1A receptors; decreases exploratory locomotor activity and increases prepulse inhibition of the acoustic startle response in mice at 30 mg/kg
Catalog No. 50-293-6972 Supplier Cayman Chemical Supplier No. 2615310 MG
An active metabolite of clozapine; originally described as an antagonist of 5-HT2C receptors (IC50 = 7.1 nM in rat choroid plexus); and later as an inverse agonist using human recombinant receptors; an antagonist at dopamine D4 receptors, an agonist at δ-opioid receptors, and a partial agonist at dopamine D2 and D3, M1 muscarinic acetylcholine, and 5-HT1A receptors; decreases exploratory locomotor activity and increases prepulse inhibition of the acoustic startle response in mice at 30 mg/kg