Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Desmethylolanzapine | 161696-76-0 | 98.0% | 298.42 | 1 ML
SDP

Catalog No. 5000451558
Encompass_Preferred

N-Desmethylolanzapine is an antipsychotic drug, also known as N-Demethylolanzapine or LY170055. Its formation is linked to the levels and activity of human liver flavin-containing monooxygenase (FMO3). This compound is utilized in the study of antipsychotic medications.

  • Used in the study of antipsychotic medications
  • Purity 98.0%
  • Molecular weight 298.42
  • CAS number 161696-76-0
  • Appears as a solid
  • Color ranges from yellow to brown
  • Soluble in DMSO

Catalog No. 50-004-51558 Supplier Medchemexpress LLC Supplier No. HYW00924710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desmethylolanzapine is an antipsychotic drug, also known as N-Demethylolanzapine or LY170055. Its formation is linked to the levels and activity of human liver flavin-containing monooxygenase (FMO3). This compound is utilized in the study of antipsychotic medications.

  • Used in the study of antipsychotic medications
  • Purity 98.0%
  • Molecular weight 298.42
  • CAS number 161696-76-0
  • Appears as a solid
  • Color ranges from yellow to brown
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.