Learn More
AdipoGen N-Desmethylthiamethoxam
SDP
Supplier:  AdipoGen CDXC0134M050
Encompass_Preferred
Chemical. CAS 171103-04-1. Formula C7H8ClN5O3S. MW 277.69. Synthetic. Neonicotinoid insecticide. Interacts with insect nicotinic acetylcholine receptors nAChRs, but is about 25 times less potent than thiamethoxam as an insecticide. Shown to be almost as active as the insecticide imidacloprid. Can be used as a reference compound.
Catalog No. 50-256-1464