Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-dibenzofuran-3-yl-5-nitrofuran-2-carboxamide | 329198-87-0 | 99.5% | 322.27 g/mol | C17H10N2O5 | 250MG
SDP

Catalog No. 5000235395
Encompass_Preferred

C-178 is a potent, selective, covalent small-molecule inhibitor of the Stimulator of Interferon Genes (STING). It covalently modifies Cys91 on mouse STING, blocking palmitoylation and downstream signaling; the compound is active in mouse models but shows little to no activity against human STING. Supplied as a research-grade compound for preclinical studies; not for human or veterinary use.

  • Covalent inhibitor of STING that binds Cys91.
  • Selective activity in mouse STING assays, not active on human STING.
  • Molecular formula C17H10N2O5 and molecular weight 322.27 g/mol.
  • High purity, typically ≥99% by HPLC.
  • Appearance: yellow to orange solid; soluble in DMSO.
  • Mechanism blocks STING palmitoylation and downstream signaling events.
  • Intended for research use only; not for clinical use.

Catalog No. 50-002-35395 Supplier Medchemexpress LLC Supplier No. HY123963250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-178 is a potent, selective, covalent small-molecule inhibitor of the Stimulator of Interferon Genes (STING). It covalently modifies Cys91 on mouse STING, blocking palmitoylation and downstream signaling; the compound is active in mouse models but shows little to no activity against human STING. Supplied as a research-grade compound for preclinical studies; not for human or veterinary use.

  • Covalent inhibitor of STING that binds Cys91.
  • Selective activity in mouse STING assays, not active on human STING.
  • Molecular formula C17H10N2O5 and molecular weight 322.27 g/mol.
  • High purity, typically ≥99% by HPLC.
  • Appearance: yellow to orange solid; soluble in DMSO.
  • Mechanism blocks STING palmitoylation and downstream signaling events.
  • Intended for research use only; not for clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.