Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(Diphenylmethylene)glycine ethyl ester, 98%

Click to view available options
Quantity:
5g
25g
69555-14-2
C17H17NO2
267.328
MFCD00010590
QUGJYNGNUBHTNS-UHFFFAOYSA-N
ethyl n-diphenylmethylene glycinate,n-diphenylmethylene glycine ethyl ester,ethyl 2-diphenylmethylene amino acetate,ethyl 2-diphenylmethyleneamino acetate,diphenylmethylene-glycine ethyl ester,ethyl 2-benzhydrylideneamino acetate,ethyl 2-diphenylmethylidene amino acetate,diphenylmethylene glycine ethylester,ethyl diphenylmethylenamino acetate,diphenylmethylene glycine ethyl ester
319508
ethyl 2-(benzhydrylideneamino)acetate
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
69555-14-2
C17H17NO2
267.328
MFCD00010590
QUGJYNGNUBHTNS-UHFFFAOYSA-N
ethyl n-diphenylmethylene glycinate,n-diphenylmethylene glycine ethyl ester,ethyl 2-diphenylmethylene amino acetate,ethyl 2-diphenylmethyleneamino acetate,diphenylmethylene-glycine ethyl ester,ethyl 2-benzhydrylideneamino acetate,ethyl 2-diphenylmethylidene amino acetate,diphenylmethylene glycine ethylester,ethyl diphenylmethylenamino acetate,diphenylmethylene glycine ethyl ester
319508
ethyl 2-(benzhydrylideneamino)acetate
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
Formula Weight 267.33
Percent Purity 98%
Melting Point 51°C to 55°C
Quantity 5g
Flash Point >110°C (230°F)
Chemical Name or Material N-(Diphenylmethylene)glycine ethyl ester