Learn More
Cayman Chemical N-Docosahexaenoyl DopamIn 10mg
SDP
Catalog No. 502893507
Encompass_Preferred
An amine conjugate of the neurotransmitter dopamine and DHA that is actively transported into the brain; reduces general locomotor activity 35-40% at 5 mg/kg; exhibits immune-modulating, anti-inflammatory activity in the range of 0.1-2.5 µM
Catalog No. 50-289-3507 Supplier Cayman Chemical Supplier No. 900139410 MG
An amine conjugate of the neurotransmitter dopamine and DHA that is actively transported into the brain; reduces general locomotor activity 35-40% at 5 mg/kg; exhibits immune-modulating, anti-inflammatory activity in the range of 0.1-2.5 µM