Learn More
Medchemexpress LLC N-docosanoyl taurine | 783284-48-0 | MFCD12912230 | 99.0% | 447.7 g/mol | C24H49NO4S | 1MG
SDP
Catalog No. 5000238894
Encompass_Preferred
N-docosanoyl taurine is a lipoamino acid (a taurine-conjugated long-chain fatty acid) reported as a discriminatory metabolite in brain-region classification. It is provided as a solid research reagent for biochemical and metabolomic studies and is characterized by CAS 783284-48-0, molecular formula C24H49NO4S, and molecular weight 447.7 g/mol.
  • Lipoamino acid with taurine conjugation.
  • Reported as a discriminatory metabolite in brain-region studies.
  • Solid research reagent with formula C24H49NO4S.
  • Molecular weight 447.7 g/mol.
  • High purity suitable for analytical and biochemical research.
  • Supplied in small research quantities; store at -20 °C.
Catalog No. 50-002-38894 Supplier Medchemexpress LLC Supplier No. HY1240011MG
N-docosanoyl taurine is a lipoamino acid (a taurine-conjugated long-chain fatty acid) reported as a discriminatory metabolite in brain-region classification. It is provided as a solid research reagent for biochemical and metabolomic studies and is characterized by CAS 783284-48-0, molecular formula C24H49NO4S, and molecular weight 447.7 g/mol.
  • Lipoamino acid with taurine conjugation.
  • Reported as a discriminatory metabolite in brain-region studies.
  • Solid research reagent with formula C24H49NO4S.
  • Molecular weight 447.7 g/mol.
  • High purity suitable for analytical and biochemical research.
  • Supplied in small research quantities; store at -20 °C.