Learn More
Medchemexpress LLC N-dodecanoyl-L-homoserine lactone | 137173-46-7 | MFCD11976890 | 98.0% | 283.41 g/mol | C16H29NO3 | 1 ML
SDP
Catalog No. 5000226319
Encompass_Preferred
N-dodecanoyl-L-homoserine lactone (C12-HSL) is a bacterial quorum sensing signaling molecule supplied as a pre-made 10 mM solution in DMSO for in vitro research applications. It serves as a standard reagent in quorum sensing and biofilm assays, provided at high purity for analytical and biological testing.
  • Prepared as a 10 mM solution in DMSO, 1 mL volume.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 283.41 g/mol; formula C16H29NO3.
  • Store aliquots in solvent at -80°C for long-term stability.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-26319 Supplier Medchemexpress LLC Supplier No. HY11869710MM 1ML
N-dodecanoyl-L-homoserine lactone (C12-HSL) is a bacterial quorum sensing signaling molecule supplied as a pre-made 10 mM solution in DMSO for in vitro research applications. It serves as a standard reagent in quorum sensing and biofilm assays, provided at high purity for analytical and biological testing.
  • Prepared as a 10 mM solution in DMSO, 1 mL volume.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 283.41 g/mol; formula C16H29NO3.
  • Store aliquots in solvent at -80°C for long-term stability.
  • Intended for research use only; not for human or diagnostic use.