Learn More
Cayman Chemical n-Dodecyl-α-D-maltoside, Purity: ≥95%, CAS: 116183-64-3, 1 g
SDP
Catalog No. 502936970
Encompass_Preferred
A nonionic detergent commonly used to solubilize integral membrane proteins and protein complexes; CMC = 0.15 mM; has been used to solubilize PSI and PSII complexes from plant thylakoid membranes, as well as intact grana
Catalog No. 50-293-6970 Supplier Cayman Chemical Supplier No. 261521 G
A nonionic detergent commonly used to solubilize integral membrane proteins and protein complexes; CMC = 0.15 mM; has been used to solubilize PSI and PSII complexes from plant thylakoid membranes, as well as intact grana