Learn More
Cayman Chemical n-Dodecyl-a-D-maltosIde 500mg
SDP
Catalog No. 502936971
Encompass_Preferred
A nonionic detergent commonly used to solubilize integral membrane proteins and protein complexes; CMC = 0.15 mM; has been used to solubilize PSI and PSII complexes from plant thylakoid membranes, as well as intact grana
Catalog No. 50-293-6971 Supplier Cayman Chemical Supplier No. 26152500 MG
Add to Cart
Add to Cart
A nonionic detergent commonly used to solubilize integral membrane proteins and protein complexes; CMC = 0.15 mM; has been used to solubilize PSI and PSII complexes from plant thylakoid membranes, as well as intact grana