Learn More
AdipoGen n-Dodecyl-alpha-D-maltoside (high purity) Chemical. CAS: 116183-64-3
SDP
Supplier:  AdipoGen AGCC10006G001
Encompass_Preferred
n-Dodecyl-alpha-D-maltoside (high purity), AdipoGen Life Sciences. Chemical. CAS: 116183-64-3. Formula: C24H46O11. MW: 510.6. Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. For the stabilization and activation of enzymes.
Catalog No. 50-101-5972