Learn More
Gold Biotechnology Inc n-Dodecyl-B-D-maltoside (DDM) 10 g
SDP
Catalog No. NC2159859
Encompass
n-Dodecyl-B-D-Maltoside, or DDM, is a maltoside based non-ionic detergent with a hydrophilic maltose head and a hydrophobic long chain alkyl tail. It is considered a gentle detergent that is more efficient than other detergents, such as CHAPS or NP-40. It has a comparatively low critical micelle concentration (CMC) of 0.17mM compared to the other maltoside based detergents.
This item is not returnable. View return policy
Catalog No. NC2159859 Supplier Gold Biotechnology Inc Supplier No. DDM10
n-Dodecyl-B-D-Maltoside, or DDM, is a maltoside based non-ionic detergent with a hydrophilic maltose head and a hydrophobic long chain alkyl tail. It is considered a gentle detergent that is more efficient than other detergents, such as CHAPS or NP-40. It has a comparatively low critical micelle concentration (CMC) of 0.17mM compared to the other maltoside based detergents.