Learn More
Research Products International Corp n-Dodecyl-B-D-maltoside, 1 Gram
SDP
Catalog No. 50213274
Encompass_Preferred
Non-ionic detergent used in membrane protein studies. Reported to increase thermal stability of lipid-free rhodopsin in solution.
Catalog No. 50-213-274 Supplier Research Products International Corp Supplier No. D120001.0
Non-ionic detergent used in membrane protein studies. Reported to increase thermal stability of lipid-free rhodopsin in solution.