Learn More
Enzo Life Sciences n-Dodecyl-beta-D-maltoside (high purity) (25g). CAS: 69227-93-6
SDP
Supplier:  Enzo Life Sciences ALX500007G025
Encompass_Preferred
Non-ionic detergent similar to n-octyl-β-D-glucopyranoside (ALX-500-001). However, increased alkyl chain length has been shown to increase thermal stability of lipid-free rhodopsin in solution. Applicable to membrane-protein solubilization studies. Alternative name: n-Dodecyl-beta-D-maltopyranoside. Purity: ≥99% (HPLC)(free of α-isomer). Solubility: 100mg/ml soluble in water and 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9341