Learn More
Gold Biotechnology Inc n Dodecyl B D maltoside DDM
SDP
Catalog No. 5000562208
Encompass_Preferred
The primary attribute of DDM is its capability to extract hydrophobic proteins while maintaining the solution-phase protein conformation DDM also allows the protein to be reformed following denaturization Most membrane-bound proteins are -helix bundles which are less robust to changes in their cellular environment DDM offers a detergent capable of preserving these proteins This is in contrast to other harsher ionic detergents that destabilize and permanently denature -helical membrane-bound proteins DDM has been used to solubilize the multidrug-transporter protein EmrE as well as the nucleoside-specific porin protein Tsx
Catalog No. 50-005-62208 Supplier Gold Biotechnology Inc Supplier No. DDM50
The primary attribute of DDM is its capability to extract hydrophobic proteins while maintaining the solution-phase protein conformation DDM also allows the protein to be reformed following denaturization Most membrane-bound proteins are -helix bundles which are less robust to changes in their cellular environment DDM offers a detergent capable of preserving these proteins This is in contrast to other harsher ionic detergents that destabilize and permanently denature -helical membrane-bound proteins DDM has been used to solubilize the multidrug-transporter protein EmrE as well as the nucleoside-specific porin protein Tsx