Learn More
AdipoGen n-Dodecyl-beta-D-maltoside
SDP
Catalog No. 502562718
Encompass_Preferred
Chemical. CAS 69227-93-6. Formula C24H46O11. MW 510.6. Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. For the stabilization and activation of enzymes.
Catalog No. 50-256-2718 Supplier AdipoGen Supplier No. AGCC10004G005
Chemical. CAS 69227-93-6. Formula C24H46O11. MW 510.6. Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. For the stabilization and activation of enzymes.