Learn More
Abcam n-Dodecyl-beta-D-thiomaltoside, non-ionic detergent, 500MG
Catalog No. 502704748
Encompass_Preferred
MW 526.6 g/mol, Purity >98%. Water soluble non-ionic detergent used in protein biochemistry for the stabilization of membrane proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4748 Supplier Abcam Supplier No. AB146298500MG
MW 526.6 g/mol, Purity >98%. Water soluble non-ionic detergent used in protein biochemistry for the stabilization of membrane proteins.
The product is subject to the following: Abcam Restricted Use Statement