Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ n-Dodecyl-β-D-maltoside, ULTROL™, ≥99%, Calbiochem™,

Catalog No. 3243551GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
250 g
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
32-435-51GM 1 g Glass Bottle
32-435-55GM 5 g Glass Bottle
32-435-525GM 25 g Glass Bottle
32-435-5100GM 100 g Glass Bottle
32-435-5250GM 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 32-435-51GM Supplier MilliporeSigma™ Supplier No. 3243551GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69227-93-6
Molecular Formula C24H46O11
Molecular Weight (g/mol) 510.621
InChI Key NLEBIOOXCVAHBD-QKMCSOCLSA-N
PubChem CID 114880
ChEBI CHEBI:43769
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.