Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ n-Dodecyl-βD-glucopyranoside, ≥99%, Calbiochem™,

Catalog No. 3243511GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-435-11GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-435-11GM Supplier MilliporeSigma™ Supplier No. 3243511GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59122-55-3
Molecular Formula C18H36O6
Molecular Weight (g/mol) 348.48
InChI Key PYIDGJJWBIBVIA-UYTYNIKBSA-N
Synonym Dodecylglucoside
PubChem CID 93321
IUPAC Name (2R,3R,4S,5S,6R)-2-dodecoxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity ≥99% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.